missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-2-propen-1-ol, 98%

Manufacturer:  Alfa Aesar™ H6050203

 View more versions of this product

Catalog No. AAH6050203



Specifications

Specifications

2-Methyl-2-propen-1-ol
113°C to 115°C
513-42-8
CH2=C(CH3)CH2OH
UN2614
methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol
CC(=C)CO
72.107
72.11
32°C (90°F)
1g
C4H8O
MFCD00004737
969226
BYDRTKVGBRTTIT-UHFFFAOYSA-N
2-methylprop-2-en-1-ol
10557
98%

Chemical Identifiers

513-42-8
72.107
BYDRTKVGBRTTIT-UHFFFAOYSA-N
10557
CC(=C)CO
C4H8O
MFCD00004737
methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol
2-methylprop-2-en-1-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: METHALLYL ALCOHOL

RTECSNumber : UD5250000

TSCA : TSCA

SDS Documents