Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-2-propanesulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2-Methyl-2-propanesulfonyl chloride
CAS 10490-22-9
Linear Formula C(CH3)3SO2Cl
Formula Weight 156.63
MDL Number MFCD16547515
UN Number UN3261
Beilstein 1748764
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6691503

 
alfa aesar™
H6691503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66915MD

 
alfa aesar™
H66915MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 156.63
Chemical Name or Material 2-Methyl-2-propanesulfonyl chloride
Physical Form Solid
Assay 97%
CAS 10490-22-9
Health Hazard 1 Danger
MDL Number MFCD16547515
UN Number UN3261
Beilstein 1748764
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 -30° to -10°C
DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Linear Formula C(CH3)3SO2Cl
Sensitivity Moisture Sensitive
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

-30° to -10°C