Please login to your online account to display your discounted pricing

Alfa Aesar™ (±)-2-Methyl-2-propanesulfinamide, 97%

$260.14 - $260.14

Chemical Identifiers

Chemical Name or Material (±)-2-Methyl-2-propanesulfinamide
CAS 146374-27-8
Molecular Formula (CH3)3CS(O)NH2
Formula Weight 121.2
MDL Number MFCD05861480
Synonym (+/-)-tert-Butyl sulfinamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2538506

 
alfa aesar™
B2538506
5g Each for $260.14
Description & Specifications

Specifications

Formula Weight 121.2
Molecular Formula (CH3)3CS(O)NH2
MDL Number MFCD05861480
Synonym (+/-)-tert-Butyl sulfinamide
Melting Point 97° to 101°C
Chemical Name or Material (±)-2-Methyl-2-propanesulfinamide
Assay 97%
CAS 146374-27-8
Quantity 5g

Applications
(±)-2-Methyl-2-propanesulfinamide is used in the synthesis of 4-sulfonyliminopiperidine and its reaction with benzyl Grignard to form an important pharmaceutical building block, 4-benzyl-4-aminopiperidne. It is also used to prepare spiro tosylaziridines via methylene insertion into the derived sulfonylimine.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.