Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-2-phenylsuccinimide, 99%

$171.77 - $656.95

Chemical Identifiers

Chemical Name or Material 2-Methyl-2-phenylsuccinimide
CAS 1497-17-2
Molecular Formula C11H11NO2
Formula Weight 189.21
MDL Number MFCD00005496
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5103803

  SDS
alfa aesar™
H5103803
1g Each for $171.77

AAH5103806

  SDS
alfa aesar™
H5103806
5g Each for $656.95
Description & Specifications

Specifications

Formula Weight 189.21
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00005496
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Assay 99%
CAS 1497-17-2
Molecular Formula C11H11NO2
Chemical Name or Material 2-Methyl-2-phenylsuccinimide
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.