missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-phenylpropylmagnesium chloride, 0.5M solution in diethyl ether, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438758000

 View more versions of this product

Catalog No. AC438758000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Methyl-2-phenylpropylmagnesium chloride
−28°C
0.76
800mL
10.16,84.77
C10H13ClMg
2-methyl-2-phenyl-propanyl magnesium chloride, 2-methyl-2-phenylpropyl magnesium chloride, 2-methyl-2-phenylpropylmagnesium chloride, 2-methyl-2-phenylpropylmagnesium chloride solution, 2-methyl-2-phenylpropylmagnesium chloride solution, 0.5 m in diethyl ether, bxkyfonhssflkr-uhfffaoysa-m, chloro 2-methyl-2-phenylpropyl magnesium, magnesium, chloro 2-methyl-2-phenylpropyl, neophyl magnesium chloride, neophylmagnesium chloride
CC(C)([CH2-])C1=CC=CC=C1.[Mg+2].[Cl-]
192.97
192.97
0.7600g/mL
AcroSeal™ Glass Bottle
Colorless to Yellow
35293-35-7,60-29-7
15.23,89.84
C6H5C(CH3)2CH2MgCl
IYOKDPFKCXZKJU-UHFFFAOYSA-M
magnesium;2-methanidylpropan-2-ylbenzene;chloride
118802
Liquid

Chemical Identifiers

35293-35-7
192.97
2-methyl-2-phenyl-propanyl magnesium chloride, 2-methyl-2-phenylpropyl magnesium chloride, 2-methyl-2-phenylpropylmagnesium chloride, 2-methyl-2-phenylpropylmagnesium chloride solution, 2-methyl-2-phenylpropylmagnesium chloride solution, 0.5 m in diethyl ether, bxkyfonhssflkr-uhfffaoysa-m, chloro 2-methyl-2-phenylpropyl magnesium, magnesium, chloro 2-methyl-2-phenylpropyl, neophyl magnesium chloride, neophylmagnesium chloride
magnesium;2-methanidylpropan-2-ylbenzene;chloride
C10H13ClMg
IYOKDPFKCXZKJU-UHFFFAOYSA-M
118802
CC(C)([CH2-])C1=CC=CC=C1.[Mg+2].[Cl-]
Safety and Handling

Safety and Handling

GHS H Statement
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Reacts violently with water.
May form explosive peroxides.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wash face,hands and any

GHS Signal Word: Danger

EINECSNumber : 252-492-3

SDS Documents