Learn More
2-Methyl-2-phenylpropanenitrile, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438250010
Specifications
2-Methyl-2-phenylpropanenitrile | |
Glass bottle | |
Colorless to Yellow | |
1195-98-8 | |
100.0 | |
C10H11N | |
PGQTYXFMSZUGOW-UHFFFAOYSA-N | |
2-methyl-2-phenylpropanenitrile | |
2797591 | |
Liquid |
Authentic | |
54°C to 56°C (1.0mmHg) | |
1g | |
96.0 | |
96% min. (GC) | |
2-methyl-2-phenyl-propanenitrile, 2-methyl-2-phenyl-propionitrile, 2-methyl-2-phenylpropionitrile, 2-phenylisobutyronitrile, a,, a-dimethyl, alpha,alpha-dimethylbenzyl cyanide, alpha,alpha-dimethylphenylacetonitrile, benzeneacetonitrile,, benzeneacetonitrile, alpha,alpha-dimethyl, benzeneacetonitrile,a,a-dimethyl, phenylisobutyronitrile | |
CC(C)(C#N)C1=CC=CC=C1 | |
145.21 | |
145.21 | |
97% |
Chemical Identifiers
1195-98-8 | |
145.21 | |
2-methyl-2-phenyl-propanenitrile, 2-methyl-2-phenyl-propionitrile, 2-methyl-2-phenylpropionitrile, 2-phenylisobutyronitrile, a,, a-dimethyl, alpha,alpha-dimethylbenzyl cyanide, alpha,alpha-dimethylphenylacetonitrile, benzeneacetonitrile,, benzeneacetonitrile, alpha,alpha-dimethyl, benzeneacetonitrile,a,a-dimethyl, phenylisobutyronitrile | |
2-methyl-2-phenylpropanenitrile |
C10H11N | |
PGQTYXFMSZUGOW-UHFFFAOYSA-N | |
2797591 | |
CC(C)(C#N)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning