Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-2-phenylacetamidoxime, 97%

Chemical Identifiers

Chemical Name or Material 2-Methyl-2-phenylacetamidoxime
Formula Weight 164.2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6617003

 
alfa aesar™
H6617003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66170MD

 
alfa aesar™
H66170MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.2
Chemical Name or Material 2-Methyl-2-phenylacetamidoxime
Physical Form Crystalline Solid
Assay 97%
Color White to pale brown
Quantity 1g