missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-pentenoic Acid, TCI America™

$29.03 - $85.56

Chemical Identifiers

CAS 3142-72-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00002655
InChI Key JJYWRQLLQAKNAD-SNAWJCMRSA-N
Synonym 2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
PubChem CID 5365909
IUPAC Name (E)-2-methylpent-2-enoic acid
SMILES CCC=C(C)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M07845G View Documents TCI America
M07845G
5g Each for $29.03
 
M078425G View Documents TCI America
M078425G
25g Each for $85.56
 
Specifications

Chemical Identifiers

3142-72-1
114.144
JJYWRQLLQAKNAD-SNAWJCMRSA-N
5365909
CCC=C(C)C(=O)O
C6H10O2
MFCD00002655
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
(E)-2-methylpent-2-enoic acid

Specifications

White-Yellow
3142-72-1
MFCD00002655
2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
CCC=C(C)C(=O)O
114.144
114.14
≥98.0% (GC,T)
5g
C6H10O2
3261
JJYWRQLLQAKNAD-SNAWJCMRSA-N
(E)-2-methylpent-2-enoic acid
5365909
Crystal-Lump at 20°C
2-Methyl-2-pentenoic Acid
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0609

TSCA : Yes