Learn More
2-Methyl-2-pentenal, (E)+(Z), 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1572936
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methyl-2-pentenal | |
623-36-9 | |
0.86 | |
31°C (87°F) | |
C6H10O | |
MFCD00006978 | |
UN1988 | |
Air Sensitive | |
IDEYZABHVQLHAF-GQCTYLIASA-N | |
(E)-2-methylpent-2-enal | |
5319754 | |
97% |
(E)+(Z) | |
-90°C | |
134°C to 136°C | |
Pleasant | |
1.4495 | |
500 g | |
506124 | |
2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530 | |
CCC=C(C)C=O | |
98.145 | |
98.15 |
Chemical Identifiers
623-36-9 | |
98.145 | |
IDEYZABHVQLHAF-GQCTYLIASA-N | |
5319754 | |
CCC=C(C)C=O |
C6H10O | |
MFCD00006978 | |
2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530 | |
(E)-2-methylpent-2-enal |
Safety and Handling
GHS H Statement
H331-H318-H226-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P333+P313-P363-P370+P378q-P501c
H226-H317-H318-H331-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, FLAMMABLE, TOXIC, N.O.S.
EINECSNumber : 210-789-5
RTECSNumber : SB2100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only