missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-pentenal, (E)+(Z), 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1572914

 View more versions of this product

Catalog No. AAA1572914



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Methyl-2-pentenal
623-36-9
0.86
31°C (87°F)
C6H10O
MFCD00006978
UN1988
Air Sensitive
IDEYZABHVQLHAF-GQCTYLIASA-N
(E)-2-methylpent-2-enal
5319754
97%
(E)+(Z)
-90°C
134°C to 136°C
Pleasant
1.4495
25 g
506124
2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530
CCC=C(C)C=O
98.145
98.15

Chemical Identifiers

623-36-9
98.145
IDEYZABHVQLHAF-GQCTYLIASA-N
5319754
CCC=C(C)C=O
C6H10O
MFCD00006978
2-methyl-2-pentenal, 2-pentenal, 2-methyl, e-2-methyl-2-pentenal, 2-methyl-3-ethylacrolein, 2-methyl-2-penten-1-al, 2-methyl-2-pentene-1-al, 2,4-dimethylcrotonaldehyde, trans-2-methyl-2-pentenal, e-2-methylpent-2-enal, unii-98qag1u530
(E)-2-methylpent-2-enal
Safety and Handling

Safety and Handling

GHS H Statement
H331-H318-H226-H335
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P311-P333+P313-P363-P370+P378q-P501c

H226-H317-H318-H331-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, FLAMMABLE, TOXIC, N.O.S.

EINECSNumber : 210-789-5

RTECSNumber : SB2100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only