Please login to your online account to display your discounted pricing

2-Methyl-2-pentanol, 98%, Acros Organics


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  347610250

 View more versions of this product

Catalog No. AC347610250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 102.18
Quantity 25mL
Color Clear colorless to yellow
Beilstein 01, 409
EINECS Number 209-681-0
TSCA TSCA
Density 0.8350g/mL
CAS Max % 100.0
Chemical Name or Material 2-Methyl-2-pentanol
Infrared Spectrum Authentic
Name Note 98%
Refractive Index 1.4090 to 1.4120 (20°C, 589nm)
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C6H14O
Linear Formula CH3(CH2)2C(CH3)2OH
MDL Number MFCD00004485
CAS 590-36-3
Synonym Dimethyl n-propyl carbinol
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Methyl-2-pentanol
Name Note 98%
CAS 590-36-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H14O
Linear Formula CH3(CH2)2C(CH3)2OH
Formula Weight 102.18
MDL Number MFCD00004485
Synonym Dimethyl n-propyl carbinol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents