Please login to your online account to display your discounted pricing

2-Methyl-2-nitropropane, 99%, ACROS Organics™

$170.53 - $170.53

Chemical Identifiers

Chemical Name or Material 2-Methyl-2-nitropropane
Name Note 99%
CAS 594-70-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5% (GC)
Assay Percent Range 99%
Molecular Formula C4H9NO2
Linear Formula (CH3)3CNO2
Formula Weight 103.12
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC182960250

  SDS
thermo fisher scientific
182960250
Glass bottle 25g Each for $170.53
Description & Specifications

Specifications

Physical Form Crystalline Powder, Crystals, or Flakes
Assay ≥98.5% (GC)
Formula Weight 103.12
Quantity 25g
Color White
EINECS Number 209-851-4
TSCA TSCA
Density 0.9500g/mL
Beilstein 01, 129
CAS Max % 100.0
Chemical Name or Material 2-Methyl-2-nitropropane
Infrared Spectrum Authentic
Name Note 99%
Refractive Index 1.3990 to 1.4030 (20°C, 589nm)
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C4H9NO2
Linear Formula (CH3)3CNO2
MDL Number MFCD00007393
CAS 594-70-7
Synonym tert-Nitrobutane
CAS Min % 98.5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.