Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-2-nitro-1,3-propanediol, 97%

$409.84 - $1,330.47

Chemical Identifiers

Chemical Name or Material 2-Methyl-2-nitro-1,3-propanediol
CAS 77-49-6
Molecular Formula C4H9NO4
Formula Weight 135.12
MDL Number MFCD00024812
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2519614

  SDS
alfa aesar™
B2519614
25g Each for $409.84

AAB2519622

  SDS
alfa aesar™
B2519622
100g Each for $1,330.47
Description & Specifications

Specifications

Formula Weight 135.12
Chemical Name or Material 2-Methyl-2-nitro-1,3-propanediol
Assay 97%
MDL Number MFCD00024812
CAS 77-49-6
Health Hazard 1 Warning
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number TY8925000
TSCA TSCA
Molecular Formula C4H9NO4
Melting Point 148° to 151°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.