Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-2-imidazoline, 97+%

$127.61 - $1,679.35

Chemical Identifiers

Chemical Name or Material 2-Methyl-2-imidazoline
CAS 534-26-9
Molecular Formula C4H8N2
Formula Weight 84.12
MDL Number MFCD00051490
Beilstein 104225
Merck Index 14,5635
Synonym Lysidine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1511314

 
alfa aesar™
A1511314
25g Each for $127.61

AAA1511322

 
alfa aesar™
A1511322
100g Each for $415.61

AAA1511336

 
alfa aesar™
A1511336
500g Each for $1,679.35
Description & Specifications

Specifications

Formula Weight 84.12
Chemical Name or Material 2-Methyl-2-imidazoline
TSCA TSCA
Assay ≥97%
Molecular Formula C4H8N2
MDL Number MFCD00051490
CAS 534-26-9
Boiling Point 197°C
Beilstein 104225
Melting Point 96° to 102°C
Merck Index 14,5635
Quantity 25g
Synonym Lysidine
Solubility Information Soluble in water, alcohol, chloroform
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number NI4804995

Applications
2-Methyl-2-imidazoline is used in the synthesis of series of N,N,N′-trisubstituted ethylenediamine derivatives. It may be used in the preparation of 1-alkyl-2-methyl-2-imidazolines, via alkylation with organic halides in the presence of phase transfer catalysis and absence of solvent.

Solubility
Soluble in water, alcohol, chloroform.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.