missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-imidazoline, 95%, ACROS Organics™

Manufacturer:  Acros Organics AC439720250

 View more versions of this product

Catalog No. AC439720250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Methyl-2-imidazoline
Colorless to Yellow or White to Yellow
534-26-9
100.0
C4H8N2
VWSLLSXLURJCDF-UHFFFAOYSA-N
2-methyl-4,5-dihydro-1H-imidazole
10798
Powder or Crystals or Needles
Authentic
25g
94.0
95%
2-methyl-2-imidazoline, 2-methylimidazoline, 1h-imidazole, 4,5-dihydro-2-methyl, methylglyoxalidine, 2-imidazoline, 2-methyl, 4,5-dihydro-2-methyl-1h-imidazole, unii-987f50e3pe, 2-methyl imidazoline, 2-methyl-4,5-dihydroimidazole, 2-methyl-20imidazolin
CC1=NCCN1
84.122
84.12
94.0 to 106.0%

Chemical Identifiers

534-26-9
84.122
2-methyl-2-imidazoline, 2-methylimidazoline, 1h-imidazole, 4,5-dihydro-2-methyl, methylglyoxalidine, 2-imidazoline, 2-methyl, 4,5-dihydro-2-methyl-1h-imidazole, unii-987f50e3pe, 2-methyl imidazoline, 2-methyl-4,5-dihydroimidazole, 2-methyl-20imidazolin
2-methyl-4,5-dihydro-1H-imidazole
C4H8N2
VWSLLSXLURJCDF-UHFFFAOYSA-N
10798
CC1=NCCN1
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Documents