Please login to your online account to display your discounted pricing

2-Methyl-2-imidazoline, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methyl-2-imidazoline
CAS 534-26-9
CAS Min % 94.0
CAS Max % 100.0
Assay 94.0 to 106.0%
Assay Percent Range 95%
Molecular Formula C4H8N2
Formula Weight 84.12
Synonym 4, 5-Dihydro-2-methyl-1H-imidazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC439720050

  SDS
thermo fisher scientific
AC439720050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC439720250

 
thermo fisher scientific
AC439720250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystals or Needles
Formula Weight 84.12
Quantity 5g
Color White to yellow or colorless to yellow
Assay 94.0 to 106.0%
CAS Min % 94.0
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Methyl-2-imidazoline
Assay Percent Range 95%
Molecular Formula C4H8N2
CAS 534-26-9
Synonym 4, 5-Dihydro-2-methyl-1H-imidazole
Infrared Spectrum Authentic
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.