missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-cyclopenten-1-one, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC270360010

 View more versions of this product

Catalog No. AC270360010



Specifications

Specifications

2-Methyl-2-cyclopenten-1-one
48°C
Glass bottle
0.979
Yellow
97%
96.0
97%
CH3C5H5(0)
07, 52
ZSBWUNDRDHVNJL-UHFFFAOYSA-N
2-methylcyclopent-2-en-1-one
14266
Liquid
0.9790g/mL
Authentic
1.4780 to 1.4800
158.0°C to 161.0°C
1g
1120-73-6
100.0
C6H8O
MFCD00012275
2-cyclopenten-1-one, 2-methyl, 2-cyclopenten-1-one, methyl, 2-cyclopenten-1-one,2-methyl, 2-methyl-2-cyclopenten-1-one, 2-methyl-2-cyclopentenone, 2-methylcyclopent-2-enone, acmc-20af73, cyclopentenone, 2-methyl, cyclopentenone, methyl, methyl-cyclopentenone
CC1=CCCC1=O
96.13
96.13
97%

Chemical Identifiers

1120-73-6
96.13
ZSBWUNDRDHVNJL-UHFFFAOYSA-N
14266
CC1=CCCC1=O
C6H8O
MFCD00012275
2-cyclopenten-1-one, 2-methyl, 2-cyclopenten-1-one, methyl, 2-cyclopenten-1-one,2-methyl, 2-methyl-2-cyclopenten-1-one, 2-methyl-2-cyclopentenone, 2-methylcyclopent-2-enone, acmc-20af73, cyclopentenone, 2-methyl, cyclopentenone, methyl, methyl-cyclopentenone
2-methylcyclopent-2-en-1-one
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep contai

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents