missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-2-butene, tech. 90%, remainder mainly 2-methyl-1-butene

Manufacturer:  Alfa Aesar™ 3185322

 View more versions of this product

Catalog No. AA3185322



Specifications

Specifications

2-Methyl-2-butene
−45° (−49°F)
1.3870
Colorless
100g
Remainder mainly 2-methyl-1-butene
MFCD00009276
14,607
BKOOMYPCSUNDGP-UHFFFAOYSA-N
2-methylbut-2-ene
10553
70.13
90%
0.662g/mL
Mild
36°C to 40°C
-134°C
513-35-9
C5H10
UN2460
2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
CC=C(C)C
70.135
CHEBI:77916
Liquid
Technical

Chemical Identifiers

513-35-9
70.135
BKOOMYPCSUNDGP-UHFFFAOYSA-N
10553
2-methylbut-2-ene
C5H10
MFCD00009276
2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
CHEBI:77916
CC=C(C)C
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H302
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Harmful if swallowed.

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-METHYL-2-BUTENE

RTECSNumber : EM7580000

TSCA : TSCA

SDS Documents