Please login to your online account to display your discounted pricing

2-Methyl-2-butene +99%, ACROS Organics™


Quantity 250mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  126492500

 View more versions of this product

Catalog No. AC126492500

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥99.0%
Formula Weight 70.13
Quantity 250mL
Color Clear colorless to very slightly yellow
Merck Index 15, 601
Beilstein 01, 211
EINECS Number 208-156-3
TSCA TSCA
Density 0.6600g/mL
CAS Max % 100.0
CAS Min % 99.0
Synonym β-Isoamylene
Health Hazard 2 GHS P Statement Harmful if swallowed. May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Suspected of causing genetic defects. Suspected of causing cancer. Causes skin irritation. May cause drowsiness or dizziness.
Packaging Glass bottle
Refractive Index 1.3860 to 1.3880 (20°C, 589nm)
Health Hazard 3 GHS P Statement IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Store in a well-ventilated place. Keep cool. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. Take precautionary measures against static discharge.
Infrared Spectrum Authentic
Health Hazard 1 Danger
Assay Percent Range 99+%
Chemical Name or Material 2-Methyl-2-butene
CAS 513-35-9
MDL Number MFCD00009276
Name Note 99+%
Linear Formula CH3CH=C(CH3)2
Molecular Formula C5H10

Specifications

Chemical Name or Material 2-Methyl-2-butene
Name Note 99+%
CAS 513-35-9
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C5H10
Linear Formula CH3CH=C(CH3)2
Formula Weight 70.13
MDL Number MFCD00009276
Synonym β-Isoamylene
Accessories

Danger

GHS P Statement IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Store in a well-ventilated place. Keep cool. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment. Take precautionary measures against static discharge.

GHS P Statement Harmful if swallowed. May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor. Suspected of causing genetic defects. Suspected of causing cancer. Causes skin irritation. May cause drowsiness or dizziness.

SDS & Safety Documents