missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-butene, 99+%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 126490250

 View more versions of this product

Catalog No. AC126490250



Specifications

Specifications

2-Methyl-2-butene
−45°C
Glass bottle
0.66
Colorless to Yellow
25mL
513-35-9
100.0
C5H10
MFCD00009276
15, 601
BKOOMYPCSUNDGP-UHFFFAOYSA-N
2-methylbut-2-ene
10553
70.13
99+%
0.6600g/mL
Authentic
1.3860 to 1.3880
35.0°C to 38.0°C
-134.0°C
99+%
99.0
99% min. (GC)
CH3CH=C(CH3)2
01, 211
1,1,2-trimethylethylene
CC=C(C)C
70.13
CHEBI:77916
Liquid

Chemical Identifiers

513-35-9
70.13
BKOOMYPCSUNDGP-UHFFFAOYSA-N
10553
2-methylbut-2-ene
C5H10
MFCD00009276
1,1,2-trimethylethylene
CHEBI:77916
CC=C(C)C
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Suspected of causing genetic defects.
Suspected o

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a well-ventilated place.
Keep cool.
Keep away from heat/sparks/open flame

GHS Signal Word: Danger

EINECSNumber : 208-156-3

TSCA : TSCA

SDS Documents