missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-2-butanol, 99%, pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 166620010

 View more versions of this product

Catalog No. AC166620010




Specifications

Specifications

2-Methyl-2-butanol
20°C
Glass Bottle
0.8
0.1% max.
Colorless
1L
75-85-4
C5H12O
MFCD00004478
15,7253
Solubility in water: 70.7g/L (25°C). Other solubilities: miscible with alcohol,ether,benzene,chloroform,,glycerol,oils and acetone
CCC(C)(C)O
88.15
88.15
99%
0.8000g/mL
Authentic
1.4040 to 1.4060
3.7 mPa.s (25°C)
102.0°C
-12.0°C
99%
98.5% min. (GC)
C2H5C(CH3)2OH
01,388
2-butanol, 2-methyl
MSXVEPNJUHWQHW-UHFFFAOYSA-N
2-methylbutan-2-ol
6405
Liquid
Pure

Chemical Identifiers

75-85-4
88.15
MSXVEPNJUHWQHW-UHFFFAOYSA-N
6405
CCC(C)(C)O
C5H12O
MFCD00004478
2-butanol, 2-methyl
2-methylbutan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful in contact with skin.
May cause drowsiness or d

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest

GHS Signal Word: Danger

EINECSNumber : 200-908-9

RTECSNumber : SC0175000

TSCA : TSCA

SDS Documents