Learn More
2-Methyl-2-butanol, 99%, pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 166620100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methyl-2-butanol | |
75-85-4 | |
Colorless | |
102.0°C | |
Authentic | |
Plastic drum | |
1.4040 to 1.4060 | |
MFCD00004478 | |
01,388 | |
15,7253 | |
Solubility in water: 70.7g/L (25°C). Other solubilities: miscible with alcohol,ether,benzene,chloroform,,glycerol,oils and acetone | |
CCC(C)(C)O | |
88.15 | |
3.7 mPa.s (25°C) | |
99% | |
Liquid |
99% | |
-12.0°C | |
0.8000g/mL | |
20°C | |
98.5% min. (GC) | |
C5H12O | |
C2H5C(CH3)2OH | |
10 L | |
0.8 | |
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol | |
MSXVEPNJUHWQHW-UHFFFAOYSA-N | |
2-methylbutan-2-ol | |
6405 | |
88.15 | |
Pure |
Chemical Identifiers
75-85-4 | |
88.15 | |
MSXVEPNJUHWQHW-UHFFFAOYSA-N | |
6405 | |
CCC(C)(C)O |
C5H12O | |
MFCD00004478 | |
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol | |
2-methylbutan-2-ol |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful in contact with skin.
May cause drowsiness or d
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest
GHS Signal Word: Danger
EINECSNumber : 200-908-9
RTECSNumber : SC0175000
TSCA : TSCA
RUO – Research Use Only