Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-2-adamantanol, 98%

$127.93 - $500.24

Chemical Identifiers

Chemical Name or Material 2-Methyl-2-adamantanol
CAS 702-98-7
Molecular Formula C11H18O
Formula Weight 166.26
MDL Number MFCD00074811
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1658806

 
alfa aesar™
A1658806
5g Each for $127.93

AAA1658814

 
alfa aesar™
A1658814
25g Each for $500.24
Description & Specifications

Specifications

Formula Weight 166.26
Melting Point 215° to 217°C
MDL Number MFCD00074811
Molecular Formula C11H18O
Assay 98%
CAS 702-98-7
Quantity 5g
Chemical Name or Material 2-Methyl-2-adamantanol