Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-2-(4-morpholinyl)propylamine, 97+%

Chemical Identifiers

Chemical Name or Material 2-Methyl-2-(4-morpholinyl)propylamine
CAS 173336-43-1
Molecular Formula C8H18N2O
Formula Weight 158.25
MDL Number MFCD01821227
UN Number UN2735
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH50308MD

  SDS
alfa aesar™
H50308MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.25
MDL Number MFCD01821227
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Assay ≥97%
Chemical Name or Material 2-Methyl-2-(4-morpholinyl)propylamine
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Health Hazard 1 Danger
CAS 173336-43-1
Quantity 250mg
UN Number UN2735
Molecular Formula C8H18N2O