Please login to your online account to display your discounted pricing

2-Methyl-1H-indol-5-amine, 97%


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  AC34875DA

 View more versions of this product

Catalog No. AC34875DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 146.19
Physical Form Crystalline Powder or Chunks
Quantity 1g
Color Brown
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H10N2
Chemical Name or Material 2-Methyl-1H-indol-5-amine
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD02093426
Packaging Amber Glass Bottle
Infrared Spectrum Authentic
CAS 7570-49-2

Specifications

Chemical Name or Material 2-Methyl-1H-indol-5-amine
CAS 7570-49-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 97%
Molecular Formula C9H10N2
Formula Weight 146.19
MDL Number MFCD02093426
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents