Please login to your online account to display your discounted pricing

2-Methyl-1H-indol-5-amine, 97%

Chemical Identifiers

Chemical Name or Material 2-Methyl-1H-indol-5-amine
CAS 7570-49-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 97%
Molecular Formula C9H10N2
Formula Weight 146.19
MDL Number MFCD02093426
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC34875DA

  SDS
fisher scientific company
AC34875DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder or Chunks
Formula Weight 146.19
Quantity 1g
Color Brown
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Methyl-1H-indol-5-amine
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C9H10N2
MDL Number MFCD02093426
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
CAS 7570-49-2
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.