missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-1-pyrroline

Manufacturer:  Alfa Aesar™ 44853AC

 View more versions of this product

Catalog No. AA44853AC



Specifications

Specifications

2-Methyl-1-pyrroline
10°C (50°F)
25mL
C5H9N
UN2733
CTSZPNIMMLSKDV-UHFFFAOYSA-N
5-methyl-3,4-dihydro-2H-pyrrole
70103
83.13
0.878g/mL
104°C to 105°C
872-32-2
MFCD00128806
2-methyl-1-pyrroline, 3,4-dihydro-5-methyl-2h-pyrrole, 1pyrroline2methyl, 2h-pyrrole, 3,4-dihydro-5-methyl, pubchem9412, 2-methyl-1-pyrroline
CC1=NCCC1
83.134
CHEBI:78856
Liquid

Chemical Identifiers

872-32-2
83.134
CTSZPNIMMLSKDV-UHFFFAOYSA-N
70103
5-methyl-3,4-dihydro-2H-pyrrole
C5H9N
MFCD00128806
2-methyl-1-pyrroline, 3,4-dihydro-5-methyl-2h-pyrrole, 1pyrroline2methyl, 2h-pyrrole, 3,4-dihydro-5-methyl, pubchem9412, 2-methyl-1-pyrroline
CHEBI:78856
CC1=NCCC1
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

SDS Documents