missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC435568000
Specifications
2-Methyl-1-propenylmagnesium bromide | |
−15°C | |
0.952 | |
800mL | |
7.53,90.8 | |
C4H7BrMg | |
Solubility in water: reacts | |
CC(=[CH-])C.[Mg+2].[Br-] | |
159.31 | |
159.31 |
0.9520g/mL | |
AcroSeal™ Glass Bottle | |
Orange to Yellow | |
38614-36-7,109-99-9 | |
9.2,92.47 | |
2-methyl propenyl magnesium bromide, 2-methyl-1-propenyl magnesium bromide, 2-methyl-1-propenylmagnesium bromide, 2-methyl-propenylmagnesium bromide, 2-methylprop-1-enyl magnesium bromide, 2-methylprop-1-enylmagnesium bromide, 2-methylpropenyl magnesium bromide, 2-methylpropenylmagnesium bromide, bkviqodysrejqh-uhfffaoysa-m, grignard reagent | |
FFUIUNZKXBJVSC-UHFFFAOYSA-M | |
magnesium;2-methylprop-1-ene;bromide | |
4456485 | |
Solution |
Chemical Identifiers
38614-36-7 | |
159.31 | |
2-methyl propenyl magnesium bromide, 2-methyl-1-propenyl magnesium bromide, 2-methyl-1-propenylmagnesium bromide, 2-methyl-propenylmagnesium bromide, 2-methylprop-1-enyl magnesium bromide, 2-methylprop-1-enylmagnesium bromide, 2-methylpropenyl magnesium bromide, 2-methylpropenylmagnesium bromide, bkviqodysrejqh-uhfffaoysa-m, grignard reagent | |
magnesium;2-methylprop-1-ene;bromide |
C4H7BrMg | |
FFUIUNZKXBJVSC-UHFFFAOYSA-M | |
4456485 | |
CC(=[CH-])C.[Mg+2].[Br-] |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Suspected of causing cancer if inhaled.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flamma
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture
GHS Signal Word: Danger