Please login to your online account to display your discounted pricing

2-Methyl-1-propanol, 99+%, spectrophotometric grade, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  167700025

 View more versions of this product

Catalog No. AC167700025

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 74.12
Assay ≥99.0%
Quantity 2.5L
Color Clear
EINECS Number 201-148-0
TSCA TSCA
RTECS Number NP9625000
Density 0.8g/mL
Merck Index 15, 5176
Beilstein 01, 373
CAS Min % 99.0
CAS Max % 100.0
Residue after Evaporation 0.0005% max.
Water 0.05% max.
Absorbance 0.01 max. at 330nm
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Synonym Isobutyl alcohol
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Methyl-1-propanol
Assay Percent Range 99+%
Name Note 99+%
Linear Formula (CH3)2CHCH2OH
Molecular Formula C4H10O
MDL Number MFCD00004740
CAS 78-83-1

Specifications

Chemical Name or Material 2-Methyl-1-propanol
Name Note 99+%
CAS 78-83-1
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C4H10O
Linear Formula (CH3)2CHCH2OH
Formula Weight 74.12
MDL Number MFCD00004740
Synonym Isobutyl alcohol
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.

SDS & Safety Documents