missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1-propanol, 99+%, Extra Pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC158280250

 View more versions of this product

Catalog No. AC158280250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Methyl-1-propanol
28°C
Metal drum
0.005% max.
11.7mbar at 20°C
0.2% max. (K.F.)
Undesignated
25L
99.0
99% min. (GC)
(CH3)2CHCH2OH
01, 373
1-hydroxymethylpropane
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
2-methylpropan-1-ol
6560
74.12
99+%
0.8000g/mL
Authentic
1.3945 to 1.3965
0.8
4 Pas (20°C)
108.0°C
-108.0°C
78-83-1
100.0
C4H10O
MFCD00004740
15, 5176
Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone
CC(C)CO
74.12
CHEBI:46645
Liquid
Extra Pure

Chemical Identifiers

78-83-1
74.12
ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
6560
2-methylpropan-1-ol
C4H10O
MFCD00004740
1-hydroxymethylpropane
CHEBI:46645
CC(C)CO
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation occurs: Get medical advice/at

GHS Signal Word: Danger

EINECSNumber : 201-148-

SDS Documents