Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-1-phenyl-1-pentanol, erythro + threo, 97%

$135.31 - $135.31

Chemical Identifiers

Chemical Name or Material 2-Methyl-1-phenyl-1-pentanol
CAS 73177-67-0
Name Note erythro + threo
Molecular Formula C12H18O
Formula Weight 178.27
MDL Number MFCD00049211
Synonym 2-Pentyl phenyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2059606

  SDS
alfa aesar™
B2059606
5g Each for $135.31
Description & Specifications

Specifications

Formula Weight 178.27
Chemical Name or Material 2-Methyl-1-phenyl-1-pentanol
Name Note erythro + threo
Assay 97%
Molecular Formula C12H18O
MDL Number MFCD00049211
CAS 73177-67-0
Synonym 2-Pentyl phenyl carbinol
Quantity 5g