Learn More
2-Methyl-1-pentene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 127400100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Methyl-1-pentene | |
763-29-1 | |
100.0 | |
Colorless | |
62.0°C | |
Authentic | |
Glass bottle | |
1.3910 to 1.9300 | |
MFCD00009405 | |
01, I, 90 | |
2-methyl-1-pentene, 2-methylpentene, 1-pentene, 2-methyl, 4-methyl-4-pentene, 2-methyl-pentene-1, unii-okc25o38mc, okc25o38mc, acmc-1bixb, 4-01-00-00841 beilstein handbook reference, c2h5ch2c ch3 =ch2 | |
WWUVJRULCWHUSA-UHFFFAOYSA-N | |
2-methylpent-1-ene | |
12986 | |
99% |
99% | |
98.5 | |
-136.0°C | |
0.6800g/mL | |
−26°C | |
98.5% min. (GC) | |
C6H12 | |
CH3CH2CH2C(CH3)=CH2 | |
10 mL | |
0.68 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether | |
CCCC(=C)C | |
84.15 | |
84.15 | |
Liquid |
Chemical Identifiers
763-29-1 | |
84.15 | |
WWUVJRULCWHUSA-UHFFFAOYSA-N | |
12986 | |
CCCC(=C)C |
C6H12 | |
MFCD00009405 | |
2-methyl-1-pentene, 2-methylpentene, 1-pentene, 2-methyl, 4-methyl-4-pentene, 2-methyl-pentene-1, unii-okc25o38mc, okc25o38mc, acmc-1bixb, 4-01-00-00841 beilstein handbook reference, c2h5ch2c ch3 =ch2 | |
2-methylpent-1-ene |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Immediately call a POISON CENTER o
GHS Signal Word: Danger
EINECSNumber : 212-108-7
RTECSNumber : SB2230000
TSCA : TSCA
RUO – Research Use Only