missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-1-pentene, 97%

Manufacturer:  Alfa Aesar™ H53373AA

 View more versions of this product

Catalog No. AAH53373AA



Description

Description

Applications
2-Methyl-1-pentene is prepared from 2-Methyl-1-pentene. 2-methyl-1-pentene is used in copolymerization of ethylene catalyzed by nonbridged half-titanocenes. The isomerization of 2-methyl-1-pentene leads to anionic activation of c-h bonds in olefins.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

2-Methyl-1-pentene
62°C
5mL
C6H12
UN2288
2-methyl-1-pentene, 2-methylpentene, 1-pentene, 2-methyl, 4-methyl-4-pentene, 2-methyl-pentene-1, unii-okc25o38mc, okc25o38mc, acmc-1bixb, 4-01-00-00841 beilstein handbook reference, c2h5ch2c ch3 =ch2
CCCC(=C)C
84.162
84.16
−27°C (−17°F)
-136°C
763-29-1
MFCD00009405
1340499
WWUVJRULCWHUSA-UHFFFAOYSA-N
2-methylpent-1-ene
12986
97%

Chemical Identifiers

763-29-1
84.162
WWUVJRULCWHUSA-UHFFFAOYSA-N
12986
CCCC(=C)C
C6H12
MFCD00009405
2-methyl-1-pentene, 2-methylpentene, 1-pentene, 2-methyl, 4-methyl-4-pentene, 2-methyl-pentene-1, unii-okc25o38mc, okc25o38mc, acmc-1bixb, 4-01-00-00841 beilstein handbook reference, c2h5ch2c ch3 =ch2
2-methylpent-1-ene
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOHEXENES

RTECSNumber : SB2230000

TSCA : TSCA

SDS Documents