missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1-(p-tolyl)-2-propanol, TCI America™

$88.61 - $268.89

Chemical Identifiers

CAS 20834-59-7
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00029997
InChI Key LABNAHQUILHURR-UHFFFAOYSA-N
Synonym 1,1-Dimethyl-2-(p-tolyl)ethanol, alpha,alpha,4-Trimethylphenethyl Alcohol
PubChem CID 88710
IUPAC Name 2-methyl-1-(4-methylphenyl)propan-2-ol
SMILES CC1=CC=C(C=C1)CC(C)(C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M201925G View Documents TCI America
M201925G
25g Each for $268.89
 
M20195G View Documents TCI America
M20195G
5g Each for $88.61
 
Specifications

Chemical Identifiers

20834-59-7
164.248
LABNAHQUILHURR-UHFFFAOYSA-N
88710
CC1=CC=C(C=C1)CC(C)(C)O
C11H16O
MFCD00029997
1,1-Dimethyl-2-(p-tolyl)ethanol, alpha,alpha,4-Trimethylphenethyl Alcohol
2-methyl-1-(4-methylphenyl)propan-2-ol

Specifications

112°C
42°C
20834-59-7
MFCD00029997
LABNAHQUILHURR-UHFFFAOYSA-N
2-methyl-1-(4-methylphenyl)propan-2-ol
88710
Crystal-Powder at 20°C
2-Methyl-1-(p-tolyl)-2-propanol
White
25g
C11H16O
1,1-Dimethyl-2-(p-tolyl)ethanol, alpha,alpha,4-Trimethylphenethyl Alcohol
CC1=CC=C(C=C1)CC(C)(C)O
164.248
164.25
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1073

TSCA : Yes