missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1-(p-tolyl)-2-propanol 98.0+%, TCI America™

Manufacturer:  Tci America M20195G

 View more versions of this product

Catalog No. M20195G



Specifications

Specifications

2-Methyl-1-(p-tolyl)-2-propanol
White
5g
C11H16O
1,1-Dimethyl-2-(p-tolyl)ethanol, alpha,alpha,4-Trimethylphenethyl Alcohol
CC1=CC=C(C=C1)CC(C)(C)O
164.248
164.25
≥98.0% (GC)
112°C
42°C
20834-59-7
MFCD00029997
LABNAHQUILHURR-UHFFFAOYSA-N
2-methyl-1-(4-methylphenyl)propan-2-ol
88710
Crystal-Powder at 20°C

Chemical Identifiers

20834-59-7
164.248
LABNAHQUILHURR-UHFFFAOYSA-N
88710
CC1=CC=C(C=C1)CC(C)(C)O
C11H16O
MFCD00029997
1,1-Dimethyl-2-(p-tolyl)ethanol, alpha,alpha,4-Trimethylphenethyl Alcohol
2-methyl-1-(4-methylphenyl)propan-2-ol
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1073

TSCA : Yes

Documents
Promotions

Promotions