missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1-nitronaphthalene, TCI America™

Manufacturer:  TCI America M0376250G

 View more versions of this product

Catalog No. M0376250G



Specifications

Specifications

2-Methyl-1-nitronaphthalene
Undesignated
250g
C11H9NO2
1-nitro-2-methylnaphthalene, naphthalene, 2-methyl-1-nitro, 2-methyl-1-nitrophthalene, unii-0ig2r1r9f7, 2-methyl-1-nitro-naphthalene, ccris 4680, methylnitronaphthalene, pubchem11253, dsstox_cid_5761, acmc-209qr4
CC1=C(C2=CC=CC=C2C=C1)[N+](=O)[O-]
187.198
187.20
≥98.0% (GC)
188°C
81°C
881-03-8
MFCD00003914
IZNWACYOILBFEG-UHFFFAOYSA-N
2-methyl-1-nitronaphthalene
13432
Crystal-Powder at 20°C

Chemical Identifiers

881-03-8
187.198
IZNWACYOILBFEG-UHFFFAOYSA-N
13432
CC1=C(C2=CC=CC=C2C=C1)[N+](=O)[O-]
C11H9NO2
MFCD00003914
1-nitro-2-methylnaphthalene, naphthalene, 2-methyl-1-nitro, 2-methyl-1-nitrophthalene, unii-0ig2r1r9f7, 2-methyl-1-nitro-naphthalene, ccris 4680, methylnitronaphthalene, pubchem11253, dsstox_cid_5761, acmc-209qr4
2-methyl-1-nitronaphthalene
Safety and Handling

Safety and Handling

TSCA : Yes

Documents