Please login to your online account to display your discounted pricing

2-Methyl-1-nitronaphthalene 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  127290250

 View more versions of this product

Catalog No. AC127290250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 187.2
Physical Form Crystalline Powder
Quantity 25g
Color Yellow to ochre
Beilstein 05, I, 267
EINECS Number 212-917-5
TSCA TSCA
RTECS Number QJ9708200
Melting Point 75° to 81°C
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
Name Note 99%
Chemical Name or Material 2-Methyl-1-nitronaphthalene
Molecular Formula C11H9NO2
Linear Formula CH3C10H6NO2
Assay Percent Range 99%
MDL Number MFCD00003914
CAS 881-03-8

Specifications

Chemical Name or Material 2-Methyl-1-nitronaphthalene
Name Note 99%
CAS 881-03-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H9NO2
Linear Formula CH3C10H6NO2
Formula Weight 187.2
MDL Number MFCD00003914