Please login to your online account to display your discounted pricing

2-Methyl-1-nitronaphthalene 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Methyl-1-nitronaphthalene
Name Note 99%
CAS 881-03-8
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C11H9NO2
Linear Formula CH3C10H6NO2
Formula Weight 187.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC127290250

  SDS
thermo fisher scientific
127290250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 187.2
Quantity 25g
Color Yellow to ochre
Beilstein 05, I, 267
EINECS Number 212-917-5
TSCA TSCA
Melting Point 75° to 81°C
RTECS Number QJ9708200
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 2-Methyl-1-nitronaphthalene
Infrared Spectrum Authentic
Name Note 99%
Assay Percent Range 99%
Molecular Formula C11H9NO2
Linear Formula CH3C10H6NO2
MDL Number MFCD00003914
CAS 881-03-8
CAS Min % 98.5