missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1-hexene, TCI America™

Manufacturer:  TCI America M05255G

 View more versions of this product

Catalog No. M05255G



Specifications

Specifications

2-Methyl-1-hexene
Colorless
6094-02-6
MFCD00009486
IRUDSQHLKGNCGF-UHFFFAOYSA-N
2-methylhex-1-ene
22468
Clear Liquid at 20°C
92°C
5g
C7H14
3295
CCCCC(=C)C
98.19
98.19
≥97.0% (GC)

Chemical Identifiers

6094-02-6
98.19
IRUDSQHLKGNCGF-UHFFFAOYSA-N
2-methylhex-1-ene
C7H14
MFCD00009486
22468
CCCCC(=C)C
Safety and Handling

Safety and Handling

TSCA : No

Documents