Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-1-hexen-3-yne, 97%

$72.32 - $72.32

Chemical Identifiers

Chemical Name or Material 2-Methyl-1-hexen-3-yne
CAS 23056-94-2
Molecular Formula C7H10
Formula Weight 94.16
MDL Number MFCD00041629
UN Number UN3295
Beilstein 1918823
Synonym Ethyl isopropenyl acetylene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2056006

  SDS
alfa aesar™
B2056006
5g Each for $72.32

AAB2056014

  SDS
alfa aesar™
B2056014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 94.16
Synonym Ethyl isopropenyl acetylene
Flash Point 4°C (39°F)
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 103° to 104°C
Quantity 5g
Assay 97%
Odor Characteristic
Recommended Storage 2° to 8°C
UN Number UN3295
Beilstein 1918823
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
Chemical Name or Material 2-Methyl-1-hexen-3-yne
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Molecular Formula C7H10
MDL Number MFCD00041629
CAS 23056-94-2
Health Hazard 1 Danger

Applications
2-Methyl-1-hexen-3-yne, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapour.

2° to 8°C