missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1-butanethiol, TCI America™

Manufacturer:  TCI America M104125G

 View more versions of this product

Catalog No. M104125G



Specifications

Specifications

2-Methyl-1-butanethiol
Yellow
1878-18-8
MFCD00004883
2-methyl-1-butanethiol, 1-butanethiol, 2-methyl, 2-methylbutyl mercaptan, 2-methylbutanethiol, fema no. 3303, 2-methyl-butanethiol, acmc-1cuce, 2-methyl-1-butane thiol, 1-butanethiol,2-methyl, 1-mercapto-2-methylbutane
CCC(C)CS
104.211
104.21
≥95.0% (GC)
119°C
25g
C5H12S
3336
WGQKBCSACFQGQY-UHFFFAOYSA-N
2-methylbutane-1-thiol
15877
Clear Liquid at 20°C

Chemical Identifiers

1878-18-8
104.211
WGQKBCSACFQGQY-UHFFFAOYSA-N
15877
CCC(C)CS
C5H12S
MFCD00004883
2-methyl-1-butanethiol, 1-butanethiol, 2-methyl, 2-methylbutyl mercaptan, 2-methylbutanethiol, fema no. 3303, 2-methyl-butanethiol, acmc-1cuce, 2-methyl-1-butane thiol, 1-butanethiol,2-methyl, 1-mercapto-2-methylbutane
2-methylbutane-1-thiol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0464

TSCA : Yes

Documents