missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1-butanethiol, TCI America™

$174.17 - $553.06

Chemical Identifiers

CAS 1878-18-8
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.211
MDL Number MFCD00004883
InChI Key WGQKBCSACFQGQY-UHFFFAOYSA-N
Synonym 2-methyl-1-butanethiol, 1-butanethiol, 2-methyl, 2-methylbutyl mercaptan, 2-methylbutanethiol, fema no. 3303, 2-methyl-butanethiol, acmc-1cuce, 2-methyl-1-butane thiol, 1-butanethiol,2-methyl, 1-mercapto-2-methylbutane
PubChem CID 15877
IUPAC Name 2-methylbutane-1-thiol
SMILES CCC(C)CS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M10415G View Documents TCI America
M10415G
5g Each for $174.17
 
M104125G View Documents TCI America
M104125G
25g Each for $553.06
 
Specifications

Chemical Identifiers

1878-18-8
104.211
WGQKBCSACFQGQY-UHFFFAOYSA-N
15877
CCC(C)CS
C5H12S
MFCD00004883
2-methyl-1-butanethiol, 1-butanethiol, 2-methyl, 2-methylbutyl mercaptan, 2-methylbutanethiol, fema no. 3303, 2-methyl-butanethiol, acmc-1cuce, 2-methyl-1-butane thiol, 1-butanethiol,2-methyl, 1-mercapto-2-methylbutane
2-methylbutane-1-thiol

Specifications

119°C
5g
C5H12S
3336
WGQKBCSACFQGQY-UHFFFAOYSA-N
2-methylbutane-1-thiol
15877
Clear Liquid at 20°C
2-Methyl-1-butanethiol
Yellow
1878-18-8
MFCD00004883
2-methyl-1-butanethiol, 1-butanethiol, 2-methyl, 2-methylbutyl mercaptan, 2-methylbutanethiol, fema no. 3303, 2-methyl-butanethiol, acmc-1cuce, 2-methyl-1-butane thiol, 1-butanethiol,2-methyl, 1-mercapto-2-methylbutane
CCC(C)CS
104.211
104.21
≥95.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0464

TSCA : Yes