missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1-butanethiol, 95%, ACROS Organics™

Manufacturer:  Acros Organics AC184400050

 View more versions of this product

Catalog No. AC184400050



Specifications

Specifications

2-Methyl-1-butanethiol
Authentic
1.4455 to 1.4475 (20°C, 589nm)
5g
1878-18-8
100.0
C5H12S
MFCD00004883
2-methyl-1-butanethiol, 1-butanethiol, 2-methyl, 2-methylbutyl mercaptan, 2-methylbutanethiol, fema no. 3303, 2-methyl-butanethiol, acmc-1cuce, 2-methyl-1-butane thiol, 1-butanethiol,2-methyl, 1-mercapto-2-methylbutane
CCC(C)CS
104.211
104.21
≥94%
0.8400g/mL
Glass bottle
Colorless
95%
94.0
95%
C2H5CH(CH3)CH2SH
01, II, 421
WGQKBCSACFQGQY-UHFFFAOYSA-N
2-methylbutane-1-thiol
15877
Liquid

Chemical Identifiers

1878-18-8
104.211
WGQKBCSACFQGQY-UHFFFAOYSA-N
15877
CCC(C)CS
C5H12S
MFCD00004883
2-methyl-1-butanethiol, 1-butanethiol, 2-methyl, 2-methylbutyl mercaptan, 2-methylbutanethiol, fema no. 3303, 2-methyl-butanethiol, acmc-1cuce, 2-methyl-1-butane thiol, 1-butanethiol,2-methyl, 1-mercapto-2-methylbutane
2-methylbutane-1-thiol
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 217-515-3

TSCA : TSCA

SDS Documents