Please login to your online account to display your discounted pricing

2-Methyl-1,5-diphenyl-1H-pyrrole-3-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Methyl-1, 5-diphenyl-1H-pyrrole-3-carboxylic acid
CAS 109812-64-8
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C18H15NO2
Formula Weight 277.32
MDL Number MFCD01567220
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP00294DA

 
fisher scientific company
SP00294DA
Amber glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SP00294EA

 
fisher scientific company
SP00294EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 277.32
Quantity 1g
Assay 97%
CAS Min % 97.0
Molecular Formula C18H15NO2
CAS 109812-64-8
Packaging Amber glass bottle
MDL Number MFCD01567220
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Methyl-1, 5-diphenyl-1H-pyrrole-3-carboxylic acid
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Assay Percent Range 97%