Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Methyl-1,4-bis(trimethylsiloxy)benzene, 98%

$99.59 - $374.04

Chemical Identifiers

Chemical Name or Material 2-Methyl-1,4-bis(trimethylsiloxy)benzene
CAS 78018-53-8
Molecular Formula C13H24O2Si2
Formula Weight 268.51
MDL Number MFCD02683083
Synonym 2,5-Bis(trimethylsiloxy)toluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2211606

 
alfa aesar™
B2211606
5g Each for $99.59

AAB2211614

 
alfa aesar™
B2211614
25g Each for $374.04
Description & Specifications

Specifications

Formula Weight 268.51
Molecular Formula C13H24O2Si2
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Methyl-1,4-bis(trimethylsiloxy)benzene
Assay 98%
MDL Number MFCD02683083
CAS 78018-53-8
Synonym 2,5-Bis(trimethylsiloxy)toluene
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.