Please login to your online account to display your discounted pricing

2-Methyl-1,3-thiazole-4-carboxylic acid, 97%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  CC05701DE

 View more versions of this product

Catalog No. CC05701DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 143.17
Quantity 5g
Assay 97%
CAS Max % 100.0
MDL Number MFCD03407332
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C5H5NO2S
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 96.5
Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carboxylic acid
CAS 35272-15-2
Assay Percent Range 97%

Specifications

Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carboxylic acid
CAS 35272-15-2
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5NO2S
Formula Weight 143.17
MDL Number MFCD03407332
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.