Please login to your online account to display your discounted pricing

2-Methyl-1,3-thiazole-4-carbothioamide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carbothioamide
CAS 174223-29-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H6N2S2
Formula Weight 158.25
MDL Number MFCD00052167
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB02802DA

 
fisher scientific company
SPB02802DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.25
Quantity 1g
Assay 97%
CAS Min % 97.0
MDL Number MFCD00052167
Assay Percent Range 97%
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carbothioamide
Packaging Amber Glass Bottle
CAS 174223-29-1
Molecular Formula C5H6N2S2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.