Please login to your online account to display your discounted pricing

(2-Methyl-1,3-thiazol-4-yl)methanol, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC05709DA

 View more versions of this product

Catalog No. CC05709DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 129.18
Quantity 1g
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H7NOS
CAS 76632-23-0
Chemical Name or Material (2-Methyl-1, 3-thiazol-4-yl)methanol
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material (2-Methyl-1, 3-thiazol-4-yl)methanol
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H7NOS
Formula Weight 129.18
Accessories

GHS Signal Word: Warning

SDS & Safety Documents