Please login to your online account to display your discounted pricing

(2-Methyl-1,3-thiazol-4-yl)methanol, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material (2-Methyl-1, 3-thiazol-4-yl)methanol
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H7NOS
Formula Weight 129.18
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CC05709DA

  SDS
fisher scientific company
CC05709DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CC05709DE

  SDS
fisher scientific company
CC05709DE
Amber Glass Bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 129.18
Quantity 1g
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H7NOS
Chemical Name or Material (2-Methyl-1, 3-thiazol-4-yl)methanol
Packaging Amber Glass Bottle
CAS 76632-23-0
Health Hazard 1 GHS Signal Word: Warning
Safety and Handling

GHS Signal Word: Warning