missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Methyl-1,3-propanediol, TCI America™

$25.97 - $50.42

Chemical Identifiers

CAS 2163-42-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00082586
InChI Key QWGRWMMWNDWRQN-UHFFFAOYSA-N
Synonym 1,3-Dihydroxy-2-methylpropane
PubChem CID 75103
IUPAC Name 2-methylpropane-1,3-diol
SMILES CC(CO)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M111425ML View Documents TCI America
M111425ML
25mL Each for $25.97
 
M1114500ML View Documents TCI America
M1114500ML
500mL Each for $50.42
 
Specifications

Chemical Identifiers

2163-42-0
90.122
QWGRWMMWNDWRQN-UHFFFAOYSA-N
75103
CC(CO)CO
C4H10O2
MFCD00082586
1,3-Dihydroxy-2-methylpropane
2-methylpropane-1,3-diol

Specifications

214°C
25mL
C4H10O2
1,3-Dihydroxy-2-methylpropane
CC(CO)CO
90.122
90.12
≥98.0% (GC)
Yellow
2163-42-0
MFCD00082586
QWGRWMMWNDWRQN-UHFFFAOYSA-N
2-methylpropane-1,3-diol
75103
Clear Liquid at 20°C
2-Methyl-1,3-propanediol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-3203

TSCA : Yes